Tlačiť

Anti-ADAMTSL4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Dodávateľ: BIOSS INC
BS-5864R-A488
BOSSBS-5864R-A488EA 1018.63 EUR
BOSSBS-5864R-A488
Anti-ADAMTSL4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Protilátky
ADAMTSL4 is a member of ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs)-like family and has seven thrombospondin type 1 repeats. The thrombospondin type 1 repeat domain is found in many proteins with diverse biological functions including cellular adhesion, angiogenesis, and patterning of the developing nervous system. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.ADAMTSL4 is involved in the positive regulation of apoptosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADAMTSL4
Clonality:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR