Tlačiť
Anti-MUC1 Rabbit Polyclonal Antibody
Dodávateľ: BIOSS INC
BS-4763R
BOSSBS-4763REA 827.16 EUR
BOSSBS-4763R
Anti-MUC1 Rabbit Polyclonal Antibody
Protilátky
MUC1 is a large cell surface mucin glycoprotein expressed by most glandular and ductal epithelial cells and some hematopoietic cell lineages. It is expressed on most secretory epithelium, including mammary gland and some hematopoietic cells. It is expressed abundantly in lactating mammary glands and overexpressed abundantly in >90% breast carcinomas and metastases. Transgenic MUC1 has been shown to associate with all four cebB receptors and localize with erbB1 (EGFR) in lactating glands. The MUC1 gene contains seven exons and produces several different alternatively spliced variants. TheMore Product Information