Tlačiť
Anti-TNIK Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Dodávateľ: BIOSS INC
BS-4168R-A488
BOSSBS-4168R-A488EA 1018.63 EUR
BOSSBS-4168R-A488
Anti-TNIK Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Protilátky
TNIK is a MSN protein kinase that interacts with both TNF receptor-associated factor 2 (TRAF2) and the adapter protein NCK. The protein has been shown to activate the c-Jun N-terminal kinase pathway when over expressed in Phoenix-A cells. TNIK has been shown to phosphorylate gelsolin, the principal intracellular and extracellular actin-severing protein, in vitro. This and evidence from mutational studies suggest that TNIK functions in the regulation of the cytoskeleton. Northern analysis indicates TNIK expression in human heart, skeletal muscle, and brain, with lower levels of expression inMore Product Information