Tlačiť

Anti-EPO Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Dodávateľ: BIOSS INC
BS-2343R-A750
BOSSBS-2343R-A750EA 1025.44 EUR
BOSSBS-2343R-A750
Anti-EPO Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Protilátky
Erythropoietin is the principal hormone involved in the regulation of erythrocyte differentiation and the maintenance of a physiological level of circulating erythrocyte mass.

Type: Primary
Antigen: EPO
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-193/193
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR