Tlačiť
Anti-FGR Rabbit Polyclonal Antibody (Cy7®)
Dodávateľ: BIOSS INC
BS-13157R-CY7
BOSSBS-13157R-CY7EA 1026.64 EUR
BOSSBS-13157R-CY7
Anti-FGR Rabbit Polyclonal Antibody (Cy7®)
Protilátky
Src is the human homolog of the v-Src gene of the rous sarcoma virus, also designated avian sarcoma virus or ASV. Src was the first proto-oncogenic non-receptor tyrosine kinase characterized in human. The Src family, which has common structural motifs, is composed of nine members in vertebrates, including Src, Yes, Fgr, Frk, Fyn, Lyn, Hck, Lck and Blk. Src-family kinases transduce signals that are involved in the control of a variety of cellular processes, including proliferation, differentiation, motility and adhesion. Src-family ki-nases contain an amino-terminal cell membrane anchorMore Product Information
Order Now
ŠPECIFIKÁCIA
Symbol antigenu FGR
Název antigenu FGR proto-oncogene, Src family tyrosine kinase
Environmentally Preferable
Synonyma antigenu p55c-fgr|c-fgr|SRC2|p58-Fgr|p55-Fgr|p58c-fgr|c-src2
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Cy7®
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Identifikační číslo genu 2268
Izotyp IgG
Imunochemie Yes
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at 4°C for 12 months
Transportní teplota 4°C