Tlačiť
Anti-CHGC Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Dodávateľ: BIOSS INC
BS-12120R-A488
BOSSBS-12120R-A488EA 999.4 EUR
BOSSBS-12120R-A488
Anti-CHGC Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Protilátky
Chromogranins (secretogranins) are acidic glycoproteins that localize within secretory granules of endocrine, neuroendocrine and neuronal tissue. Family members include chromogranin A (Chr-A); chromogranin B (Chr-B), also known as secretogranin I; chromogranin C (also known as secretogranin II or Sg II); and secretogranin III (Sg III or SCG3). High levels of Chr-A expression are characteristic of neuroendocrine tumors. Pancreastatin is a peptide derived from Chr-A which inhibits insulin secretion, exocrine pancreatic secretion and gastric acid secretion. Pancreastatin exists as two forms;More Product Information