Tlačiť
Anti-POU2F2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Dodávateľ: BIOSS INC
BS-1077R-HRP
BOSSBS-1077R-HRPEA 1037.05 EUR
BOSSBS-1077R-HRP
Anti-POU2F2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Transcription factor that specifically binds to the octamer motif (5'-ATTTGCAT-3'). Regulates transcription in a number of tissues in addition to activating immunoglobulin gene expression. Modulates transcription transactivation by NR3C1, AR and PGR. Isoform OCT2.5 activates the U2 small nuclear RNA (snRNA) promoter. Isoforms OCT2.1, OCT2.2 and OCT2.3 activate octamer-containing promoters. Isoforms OCT2.4 and OCT2.5 repress some promoters and activate others. Isoform OCT2.7 is unable to bind to the octamer motif, but can still activate the beta-casein gene promoter at low levels.