Tlačiť
Anti-H5N1 Matrix Protein 2 Rabbit Polyclonal Antibody (Cy3®)
Dodávateľ: BIOSS INC
BS-0344R-CY3
BOSSBS-0344R-CY3EA 1026.64 EUR
BOSSBS-0344R-CY3
Anti-H5N1 Matrix Protein 2 Rabbit Polyclonal Antibody (Cy3®)
Protilátky
Influenza A virus is a major public health threat. Novel influenza virus strains caused by genetic drift and viral recombination emerge periodically to which humans have little or no immunity, resulting in devastating pandemics. Influenza A can exist in a variety of animals; however it is in birds that all subtypes can be found. These subtypes are classified based on the combination of the virus coat glycoproteins hemagglutinin (HA) and neuraminidase (NA) subtypes. During 1997, an H5N1 avian influenza virus was determined to be the cause of death in 6 of 18 infected patients in Hong Kong.More Product Information