Tlačiť
Dropping pipettes
Dodávateľ: DWK LIFE SCIENCES
SBD050 SBD100 SBD250
BIBBSBD050EA 100.14 EUR
BIBBSBD050 BIBBSBD100 BIBBSBD250
Dropping pipettes
Pipety Pipety Pasteurovy
Soda-lime glass.
  • With integral ground glass cone on pipette
  • Supplied with rubber teats
  • Bottles available separately