Tlačiť
Application specific filter papers, Fritware®, SP Bel-Art
Dodávateľ: Bel-Art Products, a Part of SP
H136385116
BELAH136385116EA 288.4 EUR
BELAH136385116
Application specific filter papers, Fritware®, SP Bel-Art
Filtry Papíry filtrační Papír kvalitativní filtrační
These HDPE sheets can be cut, sawed, formed, machined and welded to fabricate special items.
  • Medium porosity style has an average pore size of 70 µm
  • Coarse porosity style averages 110 µm