Tlačiť
Bags, safety pouch, SP Bel-Art
Dodávateľ: Bel-Art Products, a Part of SP
H132340000
BELAH132340000EA 476.67 EUR
BELAH132340000
Bags, safety pouch, SP Bel-Art
Sáčky Jednorázové odpadní sáčky
Hard to pierce pouch offering protection against the danger of being cut by contaminated sharp objects. Convenient for labs, clinics and medical offices. Used with the Poxygrid® safety pouch stand for easy benchtop collection.
  • Solid paperboard pouch
  • Remains open in an oval shape by folding the bottom flaps
  • After filling, the top flaps close and lock in place for safe disposal
  • Printed with bold biohazard symbol