Tlačiť
Repetitive pipettes, Vari-Pet®, SP Bel-Art
Dodávateľ: Bel-Art Products, a Part of SP
F378950010
BELAF378950010EA 744.24 EUR
BELAF378950010
Repetitive pipettes, Vari-Pet®, SP Bel-Art
Pipety Pipety pro opakované použití
This dependable, self-filling pipetting aid is designed for speed, ease, and convenience and is offered in two models.
  • Teflon® plunger assures smooth, consistent operation
  • PP handle is ergonomically designed to permit extended use without fatigue
  • Dispensing volume can be adjusted by turning the knob at the base of the unit
  • Excellent chemical resistance because the fluids only contact the borosilicate glass and Teflon®
  • Supplied with a chrome-plated steel Luer lock fitting, complete with removable cannula for general use
  • Autoclavable