Tlačiť
Cryo storage boxes, 0 °C Cryo-Safe™ Mini Cooler, SP Bel-Art
Dodávateľ: Bel-Art Products, a Part of SP
F188460000
BELAF188460000EA 376.53 EUR
BELAF188460000
Cryo storage boxes, 0 °C Cryo-Safe™ Mini Cooler, SP Bel-Art
Krabice Chladící/Mrazící boxy/Sáčky
Polycarbonate body and clear polycarbonate lid, non-toxic gel-filled.
  • Stackable to save space
  • Molded side handles allow secure grip for transfer
  • Rubber feet prevent sliding on the lab bench
A non-toxic freezing gel is isolated within the walls of the cooler and lid, ensuring lasting low temperatures and protecting any labels or marks on the vials. The lid is indexed and marked to show tube location that matches locations printed on the cooler. Ideal for safeguarding samples in freezers with defrost cycle and fluctuating temperatures. Keep twelve 0,5More Product Information