Tlačiť

Silicic acid magnesium salt 60-120 mesh for chromatography

Dodávateľ: Thermo Fisher Scientific

Synonymá: Magnesium silicate, aktivní, Florisil

B22490.0B B22490.30
B22490.0BEA 368.52 EUR
B22490.0B B22490.30
Silicic acid magnesium salt 60-120 mesh for chromatography
Magnesium silicate

Magnesium silicate, activated, is used in polyether adsorption and decolorization effect. Also used in chromatography.
Notes: Incompatible with strong acids and hydrogen fluoride.

Vzorec: MgO₃Si
Mw: 100.38 g/mol
Skladovacia teplota: Ambient
Číslo MDL: MFCD00078249
Číslo CAS: 1343-88-0
EINECS: 215-681-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR