Tlačiť

Genomic DNA extraction kits, plant gDNA kit, Axygen® AxyPrep™ Mag

Dodávateľ: Corning

MAGPLANTGDNA10
AXYGMAGPLANTGDNA10EA 80.91 EUR
AXYGMAGPLANTGDNA10
Genomic DNA extraction kits, plant gDNA kit, Axygen® AxyPrep™ Mag
Činidla pro nukleové kyseliny Nucleic Acid Purification Kits and Reagents

Axygen® AxyPrep™ Mag Plant gDNA Kit is a magnetic bead-based purification system that allows rapid and reliable isolation of high quality gDNA from a wide variety of plant samples. Up to 96 samples (50 mg fresh tissue per sample or 15 mg of dried tissue per sample) can be processed in less than one hour. The Axygen® AxyPrep™ Mag Plant gDNA Kit eliminates polysaccharides, phenolic compounds and enzyme inhibitors with no organic extractions. This kit is adaptable for use on major liquid handling workstations for high throughput processing.


  • High quality, high yield DNA
  • Non alcohol based purification
  • Compatible with genomic downstream applications: PCR, Sanger and next generation sequencing, fragment analysis, genotyping and SNP detection, and restriction enzyme clean up

Order Now

ŠPECIFIKÁCIA

Popis Axygen® AxyPrep™ Mag Plant gDNA Kit
Environmentally Preferable
Aplikace
Target molecule DNA
Refine target molecule Total DNA
Sample type Plant samples
Method / Format Magnetic beads
Prep size Mini

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR