Tlačiť
Digital multi timer
Dodávateľ: Amarell
E920410
AMARE920410EA 21.91 EUR
AMARE920410
Digital multi timer
Časovace/Stopky Časovače digitální
Quadruple count down/count up timer with loud alarm signal.
  • Timing capacity: 99 hours, 59 minutes
  • With stand for bench top use
Informace o dodávce: Supplied with 1,5 V AAA battery.