Tlačiť

n-Hexan 97+% ACS, meets analytical specifications of Reagent Ph. Eur. for HPLC

Dodávateľ: Thermo Fisher Scientific
Nebezpečí

Synonymá: Hexyl hydride, Hexanen, Hexane

445720010 445720025
ACRO445720010EA 104.55 EUR
ACRO445720010 ACRO445720025
n-Hexan 97+% ACS, meets analytical specifications of Reagent Ph. Eur. for HPLC
n-Hexan
Vzorec: H₃C(CH₂)₄CH₃
Mw: 86.18
Teplota varu: 69 °C (760 mmHg)
Teplota tavenia: –95 °C
Hustota: 0.659 g/cm³
Teplota vzplanutia: –22 °C
Skladovacia teplota: Ambient
Číslo MDL: MFCD00009520
Číslo CAS: 110-54-3
OSN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Výsledky špecifikačného testu

Appearance Clear liquid
Refractive index 1.3750 to 1.3760 (20°C, 589 nm)
Water ≤0.1 %
Density (20/20°C) 0.659 to 0.663
GC plus methylcyclopentane)
UV at 420 nm A: ≤0.005
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %
Acidity ≤0.0002 meq/g
Sulfur compounds (as S) ≤0.005 %
Water soluble titr. acid ≤0.0003 meq/g
Thiophene Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR