Tlačiť

Hematoxylin 85%, pure, certified

Dodávateľ: Thermo Fisher Scientific
Varování

Synonymá: DcH-48, Hydroxybrazilin, (6aS,11bR)-6,6a,7,11b-Tetrahydroindeno[2,1-c]chromene-3,4,6a,9,10-pentaol, NSC 2785, Natural Black I, Hydroxybrasilin, (+)-Hematoxylin

411160250 411161000 411165000
ACRO411160250EA 181.45 EUR
ACRO411160250 ACRO411161000 ACRO411165000
Hematoxylin 85%, pure, certified
Hematoxylin
Vzorec: C₁₆H₁₄O₆
Mw: 302.28 g/mol
Teplota tavenia: 100…120 °C
Skladovacia teplota: Ambient
Číslo MDL: MFCD00078111
Číslo CAS: 517-28-2
EINECS: 208-237-3

Order Now

Výsledky špecifikačného testu

Appearance Light beige to brown Powder
Infrared spectrum Conforms
Loss on drying ≤10 % (1 g, 105°C, 3 h)
UV spectrophotometry ≥80 % (On dry basis)
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR