Tlačiť

Kyselina chlorovodíková ∼32% in water for analysis

Dodávateľ: Thermo Fisher Scientific
Nebezpečí

Synonymá: Hydrochloric acid solution, Muriatic acid

389300025
ACRO389300025EA 82.11 EUR
ACRO389300025
Kyselina chlorovodíková ∼32% in water for analysis
Kyselina chlorovodíková
Vzorec: HCl
Mw: 36.46 g/mol
Teplota varu: 57 °C (760 mmHg)
Teplota tavenia: –35 °C
Hustota: 1.16 g/cm³
Skladovacia teplota: Ambient
Číslo MDL: MFCD00011324
Číslo CAS: 7647-01-0
OSN: 1789
ADR: 8 II

Order Now

Výsledky špecifikačného testu

Appearance Clear colorless Solution
Residue after ign. (sulfated) ≤0.0005 %
Titration with NaOH ≥32 %
Bromide (Br) ≤0.005 %
Sulfite (SO3) ≤0.0001 %
Sulfate (SO4) ≤0.0001 %
Aluminium (Al) ≤1 ppm
Ammonium (NH4) ≤0.0003 %
Arsenic (As) ≤0.1 ppm
Barium (Ba) ≤0.5 ppm
Cadmium (Cd) ≤0.01 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Cobalt (Co) ≤0.01 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.2 ppm
Lead (Pb) ≤0.02 ppm
Lithium (Li) ≤0.02 ppm
Magnesium (Mg) ≤0.1 ppm
Manganese (Mn) ≤0.01 ppm
Nickel (Ni) ≤0.02 ppm
Potassium (K) ≤0.1 ppm
Sodium (Na) ≤0.5 ppm
Strontium (Sr) ≤0.02 ppm
Zinc (Zn) ≤0.05 ppm
Heavy metals (as Pb) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR