Tlačiť
Human PVRIG, Mouse IgG2a Fc Tag PVG-H5259 is expressed from human 293 cells HEK293. It contains AA Thr 41 - Asp 171 Accession Q6DKI 7-1. 1 * 1 mg
Dodávateľ: ACROBIOSYSTEMS
Human PVRIG, Mouse IgG2a Fc Tag PVG-H5259 is expressed from human 293 cells HEK293. It contains AA Thr 41 - Asp 171 Accession Q6DKI 7-1. 1 * 1 mg