Tlačiť

Avantor® ACE® C18-300, HPLC Columns, 3 µm

Dodávateľ: Avantor
ACE-211-0302EA 773.48 EUR
ACE-211-0302 ACE-211-0501 ACE-211-0502 ACE-211-0503 ACE-211-0546 ACE-211-1002 ACE-211-1003 ACE-211-1004 ACE-211-1046 ACE-211-1204 ACE-211-1502 ACE-211-1503 ACE-211-1504 ACE-211-1546 ACE-211-7503 ACE-211-0102GD ACE-211-0103GD ACE-211-10003 ACE-211-10005 ACE-211-15003 ACE-211-05003 ACE-211-05005 ACE2111000075 ACE-211-2546 ACE-211-2502
Avantor® ACE® C18-300, HPLC Columns, 3 µm
Kolony chromatografické
Avantor® ACE® C18-300 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative. These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.

  • Designed for peptide and protein separations
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle
More Product Information
Order Now

ŠPECIFIKÁCIA

Environmentally Preferable
Naplnění uhlíkem 9,0%
S navazanou skupinou Yes
Maximální tlak 275 bar
Rozsah pH 2,0 - 8,0
Povrchová oblast 100 m²/g
Kód USP L1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR