Tlačiť

Anti-Thy1.2 Rat Monoclonal Antibody (PerCP-Cyanine5.5) [clone: 30-H12]

Dodávateľ: ABNOVA
MAB15492
ABNOMAB15492EA 459.04 EUR
ABNOMAB15492
Anti-Thy1.2 Rat Monoclonal Antibody (PerCP-Cyanine5.5) [clone: 30-H12]
Protilátky
Rat monoclonal antibody raised against native mouse Thy1.2.

Type: Primary
Antigen: Thy1.2
Clonality: Monoclonal
Clone: 30-H12
Conjugation: PerCP-Cyanine5.5
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR