Tlačiť

Anti-SELE Mouse Monoclonal Antibody (PE) [clone: TEA2/1]

Dodávateľ: ABNOVA
MAB15349
ABNOMAB15349EA 739.84 EUR
ABNOMAB15349
Anti-SELE Mouse Monoclonal Antibody (PE) [clone: TEA2/1]
Protilátky
Mouse monoclonal antibody raised against full length recombinant SELE.

Type: Primary
Antigen: SELE
Clonality: Monoclonal
Clone: TEA2/1
Conjugation: PE
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR