Tlačiť

Anti-TG Mouse Monoclonal Antibody [clone: SPM517]

Dodávateľ: ABNOVA
MAB15168
ABNOMAB15168EA 597.24 EUR
ABNOMAB15168
Anti-TG Mouse Monoclonal Antibody [clone: SPM517]
Protilátky
Mouse monoclonal antibody raised against human TG.

Type: Primary
Antigen: TG
Clonality: Monoclonal
Clone: SPM517
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR