Tlačiť

Anti-PMEL Mouse Monoclonal Antibody [clone: PMEL/783]

Dodávateľ: ABNOVA
MAB14603
ABNOMAB14603EA 597.24 EUR
ABNOMAB14603
Anti-PMEL Mouse Monoclonal Antibody [clone: PMEL/783]
Protilátky
Mouse monoclonal antibody raised against full length recombinant human SILV.

Type: Primary
Antigen: PMEL
Clonality: Monoclonal
Clone: PMEL/783
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR