Tlačiť

Anti-PXK Rabbit Polyclonal Antibody

Dodávateľ: ABGENT
AP7017C
ABGEAP7017CEA 403.77 EUR
ABGEAP7017C
Anti-PXK Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PXK (PX domain containing serine/threonine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR