Tlačiť

Anti-VEGFA Rabbit Polyclonal Antibody

Dodávateľ: ABGENT
AP6290A
ABGEAP6290AEA 403.77 EUR
ABGEAP6290A
Anti-VEGFA Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: VEGFA (vascular endothelial growth factor A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR