Tlačiť

Anti-UBXN6 Rabbit Polyclonal Antibody

Dodávateľ: ABGENT
AP5085B
ABGEAP5085BEA 403.77 EUR
ABGEAP5085B
Anti-UBXN6 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: UBXN6 (UBX domain protein 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR