Tlačiť

Anti-MYT1 Rabbit Polyclonal Antibody

Dodávateľ: ABGENT
AP3173A
ABGEAP3173AEA 403.77 EUR
ABGEAP3173A
Anti-MYT1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: MYT1 (myelin transcription factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR