Tlačiť

Anti-JAK2 Rabbit Polyclonal Antibody

Dodávateľ: ABGENT
AP20711C
ABGEAP20711CEA 403.77 EUR
ABGEAP20711C
Anti-JAK2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: JAK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR