Tlačiť

Anti-BMP9 (GDF2) Rabbit Polyclonal Antibody

Dodávateľ: ABGENT
AP2064A
ABGEAP2064AEA 403.77 EUR
ABGEAP2064A
Anti-BMP9 (GDF2) Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: BMP9 (GDF2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

ŠPECIFIKÁCIA

Symbol antigenu BMP9 (GDF2)
Environmentally Preferable
Název antigenu Phosphatidylinositol-4-Phosphate 3-Kinase, Catalytic Subunit Delta
Synonyma antigenu PI3K|p110delta|PI3-kinase p110 subunit delta|Phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta isoform|PIK3CD|PtdIns-3-kinase p110
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Human,Mouse
Hostitel Rabbit
Izotyp
Western blot Yes
ELISA Yes
Imunochemie Yes
Epitop N-Terminal ( 1–50 aa)
Křížová adsorbce No
Skladovací teplota Cool
Transportní teplota Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR