Tlačiť

Anti-SAFB Rabbit Polyclonal Antibody

Dodávateľ: ABGENT
AP17362A
ABGEAP17362AEA 403.77 EUR
ABGEAP17362A
Anti-SAFB Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SAFB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

ŠPECIFIKÁCIA

Symbol antigenu SAFB
Environmentally Preferable
Název antigenu Scaffold attachment factor B
Synonyma antigenu HET|SAFB1|SAF-B1|HAP
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Human
Hostitel Rabbit
Identifikační číslo genu 6294
Izotyp
Western blot Yes
ELISA Yes
Epitop N-terminal
Imunogen This SAFB antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 65-94 amino acids from the N-terminal region of human SAFB.
Počet aminokyselin 65 to 94
Sekvence EGNKKTSKRSSKGRKC
Molekulární hmotnost 102.642 kDa
Čištění Protein A & Peptide affinity purification
Koncentrace 0.81
Skladovací pufr pH 7,4
Skladovací teplota Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
Transportní teplota 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR