Tlačiť

Anti-STAG1 Rabbit Polyclonal Antibody

Dodávateľ: ABGENT
AP16638C
ABGEAP16638CEA 403.77 EUR
ABGEAP16638C
Anti-STAG1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: STAG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

ŠPECIFIKÁCIA

Symbol antigenu STAG1
Environmentally Preferable
Název antigenu Stromal antigen 1
Synonyma antigenu SA1|SCC3A
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Human
Hostitel Rabbit
Identifikační číslo genu 10274
Izotyp
Western blot Yes
ELISA Yes
ImunoFluorescence Yes
Průtoková cytometrie Yes
Epitop internal
Imunogen This STAG1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 540-568 amino acids from the Central region of human STAG1.
Počet aminokyselin 540 to 568
Sekvence KERKTQIDDRNKLTC
Molekulární hmotnost 144.427 kDa
Čištění Protein A & Peptide affinity purification
Koncentrace 0.57
Skladovací pufr pH 7,4
Skladovací teplota Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
Transportní teplota 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR