Tlačiť

Anti-delta Sarcoglycan Goat polyclonal antibody unconjugated

Dodávateľ: ABCAM
AB115019-200
ABCAAB115019-200EA 1126.78 EUR
ABCAAB115019-200
Anti-delta Sarcoglycan Goat polyclonal antibody unconjugated
Protilátky
Type: Primary
Antigen: delta Sarcoglycan
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Rat
Order Now

ŠPECIFIKÁCIA

Název antigenu delta Sarcoglycan
Environmentally Preferable
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Human,Rat
Hostitel Goat
Izotyp IgG
Western blot Yes
Skladovací teplota Cool
Transportní teplota Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR