Tlačiť
6-ROXtra™ Fluorescence Reference Solution *25 µM for PCR Reactions*
Dodávateľ: AAT BIOQUEST
398
AATB398EA 491.89 EUR
AATB398
6-ROXtra™ Fluorescence Reference Solution *25 µM for PCR Reactions*
Test Stanovení pro qPCR
6-ROX is predominately used as a reference dye for performing PCR detections.
  • Multiple bioconjugation applications
  • 95% purity
Pozor: For research use only.
Order Now
ŠPECIFIKÁCIA
Čistota 95%
Environmentally Preferable