Tlačiť

6-NED maleimide fluorescent probe

Dodávateľ: AAT BIOQUEST
Varování

218
AATB218EA 469.46 EUR
AATB218
6-NED maleimide fluorescent probe
6-NED maleimide

5'-Labeled primer pairs include a fluorescently labeled oligo with a choice of dye on the 5' end, along with an unlabeled oligo, in each pair.


  • Thiol-reactive probe

Mw: 669.82 g/mol
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64179883d32d584d07656f1acf6d9bc104be22d8&vid=7bdb69df3e128721b9759360b75928f945688af5&prid=web&clid=SAPDAM

Order Now

ŠPECIFIKÁCIA

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR