Tlačiť
StrandBrite™ Green Fluorimetric RNA Quantitation Kit *High Selectivity*
Dodávateľ: AAT BIOQUEST
17657
AATB17657EA 574 EUR
AATB17657
StrandBrite™ Green Fluorimetric RNA Quantitation Kit *High Selectivity*
Test Nucleic Acid Assays
The major challenge to analyse RNA in live cells is the interferences caused by DNA.
  • Highly sensitive RNA detection
  • Robust protocol
  • Multiple applications
Pozor: For research use only.