Tlačiť
ddGTP [2',3'-Dideoxyguanosine-5'-triphosphate] *10 mM in ddH₂O* (1 µmole)
Dodávateľ: AAT BIOQUEST
17210
AATB17210EA 312.04 EUR
AATB17210
ddGTP [2',3'-Dideoxyguanosine-5'-triphosphate] *10 mM in ddH₂O* (1 µmole)
Činidla pro nukleové kyseliny Činidla Sekvenační
Sanger sequencing, also known as the chain termination method, is a technique for DNA sequencing based upon the selective incorporation of chain-terminating dideoxynucleotides (ddNTPs) by DNA polymerase.
  • In-house quality control
  • Useful for sanger sequencing applications
Pozor: For research use only.