Tlačiť
Ac-LKKR-AMC
Dodávateľ: AAT BIOQUEST
13446
AATB13446EA 296.01 EUR
AATB13446
Ac-LKKR-AMC
Ac-LKKR-AMC
Ac-Leu-Lys-Lys-Arg-AMC is a fluorogenic aminopeptidase and endopeptidase substrate.
  • Stable for 12 months
Quality tested in-house
Pozor: For Research Use Only