Tlačiť

G418 disulfát solution

Dodávateľ: PanReac AppliChem

Synonymá: G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate

A6798.0020 A6798.0050
A6798.0020EA 179.85 EUR
A6798.0020 A6798.0050
G418 disulfát solution
G418 disulfát

G418 disulphate supplied as a ready-to-use sterile solution.


Concentration: 50 mg/ml

Vzorec: C₂₀H₄₄N₄O₁₈S₂
Mw: 692.72
Číslo MDL: MFCD00058314
Číslo CAS: 108321-42-2

Order Now

ŠPECIFIKÁCIA

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR