Tlačiť

(±)-3-Octanol ≥97%

Dodávateľ: Thermo Fisher Scientific
Varování

Synonymá: 3-Octanol, (±)-Ethyl pentyl carbinol, (±)-3-Hydroxy octane, (±)-Pentyl ethyl carbinol, DL-3-Octanol

A14488.18 A14488.30
A14488.18EA 88.92 EUR
A14488.18 A14488.30
(±)-3-Octanol ≥97%
(±)-3-Octanol

Purity: 97% CAS No.: 589-98-0 Molecular Formula: C8H18O Formula Weight: 130.23 Flash Point: 68°(154°F) Refractive Index: 1.4270 Odour: Pleasant Melting Point: -45° Boiling Point: 175-176° Beilstein Reference: 1719310 MDL No.: MFCD00004590

Vzorec: C₈H₁₈O
Mw: 130.23
Teplota varu: 175…176 °C
Teplota tavenia: –45 °C
Hustota: 0.820
Teplota vzplanutia: 68 °C (154 °F)
Skladovacia teplota: Ambient
Číslo MDL: MFCD00004590
Číslo CAS: 589-98-0
EINECS: 209-667-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR