Tlačiť

Glycylglycylglycine (Gly-Gly-Gly)

Dodávateľ: Thermo Fisher Scientific

A13778.03 A13778.06 A13778.14
A13778.03EA 35.77 EUR
A13778.03 A13778.06 A13778.14
Glycylglycylglycine (Gly-Gly-Gly)
Proteiny

  • EINECS: 209-122-0
  • Melting point: ∼240 °C (dec.)
  • Merck index_V14: 14,6579


Purity: 98% CAS No.: 556-33-2 Molecular Formula: H2NCH2(CONHCH2)2CO2H Formula Weight: 191.14 Melting Point: ca 240° dec. Beilstein Reference: 1711130 Merck Reference: 14,6579 MDL No.: MFCD00036223

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR