Tlačiť

n-Hexan 95,0 GC, corrected for water content HPLC/UHPLC, Spectrophotometry, J.T.Baker®

Dodávateľ: Avantor
Nebezpečí

Synonymá: Hexyl hydride, Hexanen, Hexane

9304-03EA 0 EUR
9304-03
n-Hexan 95,0 GC, corrected for water content HPLC/UHPLC, Spectrophotometry, J.T.Baker®
n-Hexan

For use in liquid chromatography and spectrophotometry. UV cutoff 192 nm. Lot analysis on label.

Vzorec: H₃C(CH₂)₄CH₃
Mw: 86.18
Teplota varu: 69
Teplota tavenia: –94.3 °C
Hustota: 0.659 g/cm³ (20 °C)
Teplota vzplanutia: –22 °C
Skladovacia teplota: Ambient
Číslo MDL: MFCD00009520
Číslo CAS: 110-54-3
EINECS: 203-777-6
OSN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Výsledky špecifikačného testu

For Laboratory,Research,or Manufacturing Use = 0.005
Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm ≤0.005
Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm ≤0.005
Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm ≤0.008
Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm ≤0.08
Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm ≤0.2
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤192
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission ≤0.3 ppb
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤1.0 ppb
Assay (Total Saturated C₆ Isomers) (by GC, corrected for water) ≥99.5 %
Assay (as n-Hexane) (by GC, corrected for water) ≥95.0 %
Residue after Evaporation ≤2.0 ppm
Substances Darkened by H₂SO₄ Passes Test
Water (by KF, coulometric) ≤0.01 %
Filtered through a 0.2 micron filter.
Packaged under Nitrogen.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR