Tlačiť
VWR® ARISTAR®, ICP Multi-Element Quality Control Standard 10 mg/L, 22 Elements in Nitric Acid 5%
Dodávateľ: VWR Chemicals
88724.150EA 609.25 EUR
88724.150
VWR® ARISTAR®, ICP Multi-Element Quality Control Standard 10 mg/L, 22 Elements in Nitric Acid 5%
Standardy Standardy kationické Standardy víceprvkové
22 elements 10 mg/L: As, Ba, Be, Cd, Co, Cr, Cu, Fe, Al, Mn, Mo, Ni, Pb, Sb, Se, Sn, Ti, Tl, V, U, Te, Zn.
Certifikácie: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Informace o dodávce: Supplied with certificate of analysis.