Tlačiť
VWR®, Turbidity standards, formazin
Dodávateľ: VWR Chemicals
88231.180EA 71.78 EUR
88231.180 88231.270 88232.180 88232.270 88233.180 88233.270 88234.180 88234.270 88235.180 88235.270 88236.180 88236.270 88237.180 88237.270 88238.180 88238.270 88240.180 88240.270 88241.180 88241.270 88244.180 88244.270 88245.180 88245.270 88246.180 88246.270 88247.180 88247.270
VWR®, Turbidity standards, formazin
Standardy Standardy turbidimetrické
All turbidity standard solutions are Certified Reference Materials.
This standard also meets the requirements of the European Pharmacopoeia on a “2.2.1. Clarity and degree of opalescence of liquids” and ISO 7027-1 “Water quality – Determination of turbidity – part 1: Quantitative methods".
Certifikácie: Produced and calibrated under a quality system that is:
- ISO 9001 certified
- Accredited according to ISO/IEC 17025 - Testing
- Accredited according to ISO 17034 - Reference Material Producer