Tlačiť

L(+)-Kyselina askorbová Reag. Ph. Eur. 1008401

Dodávateľ: VWR Chemicals
Nebezpečí

Synonymá: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum

87782.180EA 73.3 EUR
87782.180
L(+)-Kyselina askorbová Reag. Ph. Eur. 1008401
L(+)-Kyselina askorbová

0.1% ascorbic acid in N,N-dimethylformamide


Objednávacie informácie: Normally manufactured to order. Please check with customer services.


Informace o dodávce: Supplied with certificate of analysis.

Vzorec: C₆H₈O₆
Mw: 176.13
Teplota varu: 553
Číslo MDL: MFCD00064328
Číslo CAS: 50-81-7
OSN: 2265
ADR: 3 III
Merck Index: 13,00837
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c03499908e269560d680b0d7c4651f97a367aa0&vid=73a720d6900e713a98164022273a8afa6114ad7b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR