Tlačiť
Chlorine, sensitive test strips, Dosatest
Dodávateľ: VWR Chemicals
85712.601EA 36.81 EUR
85712.601
Chlorine, sensitive test strips, Dosatest
Kity pro environmentální testy
Semi-quantitative test strips for the determination of chlorine.
  • Ideal for rapid and easy analysis of chlorine directly on-site
  • No maintenance or calibration is required
Colour change: Yellow to violet
Informace o dodávce: Delivered as 100 test strips in a tube, additional regaents, instruction leaflet in a box