Tlačiť
Nitrite test strips, Dosatest
Dodávateľ: VWR Chemicals
85705.601EA 38.45 EUR
85705.601
Nitrite test strips, Dosatest
Kity pro environmentální testy
Semi-quantitative test strips for the determination of nitrite.
  • Ideal for rapid and easy analysis of nitrite directly on-site
  • No maintenance or calibration is required
Colour change: Yellow to red
Informace o dodávce: Delivered as 100 test strips in a tube, instruction leaflet in a box
Pozor: Strongly acidic solutions must be diluted with dilute sodium hydroxide and strongly alkaline samples with dilute sulphuric acid to pH 4 to 6.