Tlačiť
VWR®, n-Heptane, secondary reference standard for GC, PESTINORM®
Dodávateľ: VWR Chemicals
85667.180EA 262.77 EUR
85667.180
VWR®, n-Heptane, secondary reference standard for GC, PESTINORM®
Standardy Standardy pro chromatografii
A secondary GC solvent reference standard for various chromatographic and analytical applications.
  • Purity and the specific impurity profile is determined by GC-MS and reported in the CoA
  • Traceability data to specific current lot of primary standard
  • Completely synthetic and typically over 99,9% pure by GC-FID