Tlačiť
VWR® PESTINORM®, Ethanol, Secondary Reference Standard for GC
Dodávateľ: VWR Chemicals
85665.180EA 262.77 EUR
85665.180
VWR® PESTINORM®, Ethanol, Secondary Reference Standard for GC
Standardy Standardy pro chromatografii
A secondary GC solvent reference standard for various chromatographic and analytical applications.
  • Purity and the specific impurity profile is determined by GC-MS and reported in the CoA
  • Traceability data to specific current lot of primary standard
  • Completely synthetic and typically over 99,9% pure by GC-FID